Learn More
CD45RA Mouse anti-Rat, Brilliant Violet 421, Clone: OX-33, BD Optibuild™
Shop All BD Biosciences ProductsDescription
The OX-33 antibody specifically recognizes a high-molecular-weight form of CD45 found only on B lymphocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
Specifications
| Antigen | CD45RA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OX-33 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 421 |
| Description | Ptprc; Lca; Leucocyte common antigen |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Leukocyte common antigen purified from rat splenocytes |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.