Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chorismic acid from Enterobacter aerogenes

Catalog No. C176110MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C176110MG 10 mg
C17615MG 5 mg
C1761100MG 100 mg
C176125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. C176110MG Supplier Sigma Aldrich Supplier No. C176110MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Intermediate in the biosynthesis of aromatic amino acids via the shikimate pathway.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • C1761.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 617-12-9
Molecular Formula C10H10O6
Linear Formula C10H10O6
MDL Number MFCD00135935
Quantity 10 mg
Synonym trans-3-([1-Carboxyethenyl]oxy)-4-hydroxy-1,5-cyclohexadiene-1-carboxylic acid
Molecular Weight (g/mol) 226.18
Percent Purity ≥80%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.