Learn More
Description
- Chloroacetonitrile can be used:
- For the synthesis of ionic liquids containing nitrile functionalized imidazolium salts
- For the synthesis of cyanomethyl dodecyl trithiocarbonate, a reversible addition-fragmentation chain transfer (raft) agent
- As an alkylating reagent for the synthesis of derivatives of triazolyl thiazoles to be used as anti-tuberculosis agents
- As a reagent in photo meerwein addition reaction for amino-arylation of alkenes
- As an esterifying agent in the synthesis of a cytotoxic natural product: apicularen a
Specifications
Specifications
| CAS | 107-14-2 |
| Density | 1.193 g/mL (at 25°C (literature)) |
| Boiling Point | 124°C to 126°C (lit.) |
| Molecular Formula | C2H2ClN |
| Refractive Index | n20/D 1.422 (literature) |
| Linear Formula | ClCH2CN |
| MDL Number | MFCD00001885 |
| Quantity | 500 g |
| Molecular Weight (g/mol) | 75.5 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.