Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl cyanoacetate

Catalog No. E18425250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E18425250G 250 g
E184255G 5 g
E184251KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E18425250G Supplier Sigma Aldrich Supplier No. E18425250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl cyanoacetate

Specifications

CAS 105-56-6
Melting Point -22°C (lit.)
Density 1.063 g/mL (at 25°C (literature))
Boiling Point 208°C to 210°C (lit.)
Molecular Formula C5H7NO2
Linear Formula NCCH2COOC2H5
Refractive Index n20/D 1.418 (literature)
Quantity 250 g
MDL Number MFCD00001940
Molecular Weight (g/mol) 113.11
Percent Purity ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.