Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl Diazoacetate

Catalog No. E222011KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
20 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E222011KG 1 kg
E222015G 5 g
E22201100G 100 g
E2220120G 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. E222011KG Supplier Sigma Aldrich Supplier No. E222011KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl Diazoacetate

Specifications

CAS 623-73-4
Melting Point -22°C
Density 1.085 g/mL at 25°C
Boiling Point 140°C to 141°C (720 mmHg, lit.)
Molecular Formula C4H6N2O2
Refractive Index n20/D 1.46
Linear Formula N=N=CHCOOC2H5
Quantity 1 kg
MDL Number MFCD00001989
Synonym DAAE
Molecular Weight (g/mol) 114.1 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.