Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl isocyanate

Catalog No. E333005G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E333005G 5 g
E33300500G 500 g
E3330025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E333005G Supplier Sigma Aldrich Supplier No. E333005G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl isocyanate may be used to generate thermoresponsiveness in polyglycidol via partial hydrophobic modification
  • The poly(glycidol-co-ethyl glycidyl carbamate) thus formed can undergo photocrosslinking in the presence of a photosensitizer to form thermoresponsive gels
  • It can also be used in the synthesis of 4,5,6,7-tetrahydrobenzo[1,2-d]thiazole-2,6-diamine derivatives that are potent DNA gyrase inhibitors

Specifications

CAS 109-90-0
Density 0.898 g/mL (at 25°C (literature))
Boiling Point 60°C (lit.)
Molecular Formula C3H5NO
Refractive Index n20/D 1.381 (literature)
Linear Formula C2H5NCO
MDL Number MFCD00002042
Quantity 5 g
Molecular Weight (g/mol) 71.08
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.