Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ FISH Tag™ DNA Kits with Alexa Fluor™ Dyes

Catalog No. F32947
Encompass_Preferred
Change view
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647
Alexa Fluor 555
Alexa Fluor 594
Color:
Green
Multi-color: Green, Orange, Red, Far-Red
Orange
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Label or Dye Color
F32947 Alexa Fluor 488 Green
F32948 Alexa Fluor 555 Orange
F32949 Alexa Fluor 594 Red
F32951 Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647 Multi-color: Green, Orange, Red, Far-Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. F32947 Supplier Molecular Probes™ Supplier No. F32947
Add to Cart
Add to Cart

FISH Tag DNA kits provide a complete solution for fluorescence in situ hybridization (FISH) applications. Choose from kits using Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, or multicolor dyes for probe synthesis, purification, and labeling of specimens for imaging.

Choose from three, single-color FISH Tag DNA kits with Alexa Fluor dye options (green, orange, or red). The FISH DNA multicolor kit contains four spectrally distinct dyes (green, orange, red, and deep-red) to allow simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and in situ analysis of mitochondrial DNA localization and content. All kits include 16 reagents and come with a comprehensive, detailed protocol to help ensure your success.

FISH Tag™ DNA Kits provide a complete workflow solution for fluorescence in situ hybridization (FISH) applications. Each kit provides all of the reagents needed for probe synthesis, labeling, and purification, as well as for imaging the labeled specimen. Choose from FISH Tag™ DNA Kits in three Alexa Fluor™ colors or get the FISH Tag™ DNA Multicolor Kit (Cat. No. F32951), which contains four spectrally distinct dyes to allow simultaneous localization of multiple sequence-specific DNA targets in chromosomal spreads and for in situ analysis of mitochondrial DNA localization and content. All kits include 16 reagents and come with a comprehensive, detailed protocol to help ensure your success.

FISH Tag™ Kit Specifications:

  • Dye (Ex/Em): Alexa Fluor™ 594 (590/615 nm)
  • Comprehensive kit includes 16 reagents
  • Comes with detailed protocol to help ensure your success

Complete Workflow Solution for FISH

FISH Tag™ Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. PureLink™ nucleic acid purification technology is used for fast and efficient purification of the labeled probe, and SlowFade™ Gold antifade reagent is added to preserve the fluorescent signal during imaging.

More Options for Nucleic Acid Labeling

To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Color Green
Detection Method Fluorescence
Format Kit
Label or Dye Alexa Fluor 488
Final Product Type Probes (Labeled DNA), cDNA (Labeled)
Sample Type DNA, Tissue (FFPE), Tissue (CRYO), Cell Culture Cells
Labeling Target DNA (General), cDNA
Excitation/Emission 492/520 nm
For Use With (Application) FISH Tag, fluorescent microscopy, DNA In situ hybridization
No. of Reactions 10 Reactions
Product Line Alexa Fluor, FISH Tag
Product Type DNA In Situ Hybridization Assay Kit
Quantity 1 Kit
Target DNA
Target Specificity Not Target-Specific
Shipping Condition Dry Ice
Show More Show Less
For imaging, what are the limitations of FISH probes?

FISH probes are generally from 300 to 1500 bp in length. To adequately image a gene or other target, the FISH probes must cover a contiguous length of ˜10 kbp. Shorter strand lengths or low-abundance DNA or RNA targets may be detected with the use of fluorescent amplification methods, such as tyramide signal amplification (TSA). For more information using TSA to amplify FISH detection, see this manual (http://tools.thermofisher.com/content/sfs/manuals/mp20912.pdf).

When should I use an RNA FISH probe versus a DNA FISH probe?

RNA FISH probes are the best option for hybridization to mRNA, rRNA, or other RNA targets. DNA FISH probes can also be used to detect RNA targets, but they do not hybridize as well to mRNA and other RNA target molecules. DNA FISH probes are routinely used to detect gene loci on chromosomes. RNA FISH probes can also be used to detect genes on chromosomes, but DNA FISH probes are more robust for this application.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.