Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GeneRuler 100 bp Plus DNA Ladder
GSA_VAGREENER_CHOICE

Catalog No. FERSM0321
Change view
Click to view available options
Quantity:
5 x 50 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERSM0321 50 μg
FERSM0322 5 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERSM0321 Supplier Thermo Scientific™ Supplier No. SM0321
Only null left
Add to Cart
Edge
Add to Cart

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

GeneRuler 100bp Plus DNA Ladder 100 to 3000bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 100bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).

The ladder is supplied with 6X DNA Loading Dye.

The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 100 bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Size Range 100 to 3000 bp
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Quantity 50 μg
Product Line GeneRuler
Product Type DNA Ladder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.