Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium glyoxylate monohydrate

Catalog No. G45021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G45021G 1 g
G45025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G45021G Supplier Sigma Aldrich Supplier No. G45021G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium glyoxylate monohydrate

Specifications

CAS 918149-31-2
Quantity 1 g
Molecular Formula C2HNaO3 -+ H2O
Linear Formula HC(O)COONa -+ H2O
MDL Number MFCD00150939
Synonym Glyoxylic acid sodium salt monohydrate; Oxoethanoic acid sodium salt
Molecular Weight (g/mol) 114.03
Percent Purity ≥93%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.