Learn More
LIFE TECHNOLOGIES SAFE IMAGER 2.0 BLUE-LIGHT TRA
Frequently Asked Questions (FAQs)
Similarly to ethidium bromide, SYBR Safe DNA Gel Stain runs in the opposite direction of the migrating DNA. This has no practical effect on the use of gels cast with SYBR Safe DNA Gel Stain, as only the very bottom of the gel will have a lower concentration of stain. This effect can be partially counteracted by staining the gel with SYBR Safe DNA Gel Stain after electrophoresis. Solutions of dye should not be added to the running buffer as this can cause breakdown of the dye at the electrodes and release toxic volatile compounds into the air.
DNA stained with SYBR Safe DNA Gel Stain can be viewed using a blue-light transilluminator such as our Safe Imager 2.0 Blue-Light Transilluminator or a standard UV transilluminator. If you plan to use the DNA for cloning, avoid exposing DNA stained with SYBR Safe DNA Gel Stain to UV light.
Please see the instrument specifications below:
- Instrument dimensions: 195 - 325 - 65 mm (11.6 - 12.8 - 2.6 in)
- Viewing surface dimensions: 190 - 190 mm (7.5 - 7.5 in)
- Light source: light emitting diodes (LED) producing a narrow emission peak centered at ~470 nm
- LED life: 50,000 hours
- Complies with the European Community Safety requirements
- Contains Class 1 LED products
- Included accessories: amber filter unit, viewing glasses, and international power cord
The E-Gel Safe Imager Real-Time Transilluminator docks onto the E-Gel iBase Power System and allows real-time imaging of the migration of DNA/RNA in E-Gel Agarose Gels containing SYBR Safe stain. The Safe Imager 2.0 Blue-Light Transilluminator is an independent system designed for viewing stained gels on the benchtop.
Yes, SYBR Safe stain is easily removed from nucleic acids by ethanol precipitation or by the ethanol wash steps used for purification spin columns.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.