Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isonicotinamide Non-distribution product as customer accommodation.  DFS Item

Catalog No. I1745125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1745125G 25 g
I17451100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1745125G Supplier Sigma Aldrich Supplier No. I1745125G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isonicotinamide

Specifications

CAS 1453-82-3
Melting Point 155°C to 157°C (lit.)
Molecular Formula C6H6N2O
Linear Formula C6H6N2O
MDL Number MFCD00006432
Quantity 25 g
Synonym Pyridine-4-carboxylic acid amide
Molecular Weight (g/mol) 122.12
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.