Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isoquinoline

Catalog No. I28208500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I28208500G 500 g
I282085G 5 g
I28208100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I28208500G Supplier Sigma Aldrich Supplier No. I28208500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isoquinoline

Specifications

CAS 119-65-3
Melting Point 26°C to 28°C (lit.)
Density 1.099 g/mL (at 25°C (literature))
Boiling Point 242°C to 243°C (lit.)
Molecular Formula C9H7N
Refractive Index n20/D 1.623 (literature)
Linear Formula C9H7N
MDL Number MFCD00006898
Quantity 500 g
Molecular Weight (g/mol) 129.16
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.