Learn More
MilliporeSigma Milli-Q™ IQ 7003/05 Ultrapure Water Purification Kit
Catalog No. IQ700XPKIT Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
IQ700XPKIT Tap
1 options
Catalog No. IQ700XPKIT Supplier MilliporeSigma Supplier No. IQ700XPKIT
Add to Cart
Add to Cart
For Milli-Q™ IQ 7003 and 7005 ultrapure water systems. This product replaces IQ700XPKT1.
  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability
  • Supplied with one drain cap per cartridge, to remove excess water and reduce the weight of exhausted cartridges at disposal
  • 63% of the parts suppliers for the IPAK Gard™ cartridge and 78% of the parts suppliers for IPAK Quanta™ cartridge included in this kit are ISO 14001 environmental management system-certified
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Purification Method Ultrapure
Resin Type Ion Exchange Resin
Type Ultrapure Water Purification Kit