Learn More
MilliporeSigma Milli-Q™ IX 7003/05 Water Purification Kit
Catalog No. IX700XPKIT Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
IX700XPKIT Tap
1 options
Catalog No. IX700XPKIT Supplier MilliporeSigma Supplier No. IX700XPKIT
Add to Cart
Add to Cart
For Milli-Q™ IX 7003/05 pure water systems. Protects RO membranes and stored pure water quality. This product replaces IX700XPKT1.
This water purification kit includes:
  • One IPAK Gard™ pretreatment cartridge composed of a pleated filter for the removal of particulates, minerals and colloids and a carbon block for the removal of free chlorine.
  • This protects the reverse osmosis (RO) membrane and improves system performance.
  • The tank vent filter protects stored pure (type 2) water from airborne contaminants.
  • It contains activated carbon, soda lime and a membrane filter to prevent impure air from entering the tank.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Type Water Purification Kit