Learn More
MilliporeSigma Milli-Q™ IX 7010/15 Water Purification Kit
Catalog No. IX70XXPKIT Shop All MilliporeSigma Products
Change view
Click to view available options
Feedwater Source:
Tap
Catalog No. Feedwater Source
IX70XXPKIT Tap
1 options
Catalog No. IX70XXPKIT Supplier MilliporeSigma Supplier No. IX70XXPKIT
Add to Cart
Add to Cart
For Milli-Q™ IX 7010/15 pure water systems. Protects RO membranes and stored pure water quality. This product replaces IX70XXPKT1.
  • e-Sure tags enable RFID connection with the system, for seamless consumable status monitoring, data management and full traceability.
  • Supplied with one drain cap per cartridge to remove excess water and reduce the weight of exhausted the cartridges at disposal.
  • 63% of the parts suppliers for the IPAK Gard™ cartridge are ISO 14001 environmental management system-certified.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Tap
Type Water Purification Kit