Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl isonicotinate

Catalog No. M52950100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M52950100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M52950100G Supplier Sigma Aldrich Supplier No. M52950100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl isonicotinate

Specifications

Melting Point 8°C to 8.5°C (lit.)
Density 1.161 g/mL (at 25°C (literature))
Boiling Point 207°C to 209°C (lit.)
Molecular Formula C7H7NO2
Refractive Index n20/D 1.512 (literature)
Linear Formula C7H7NO2
MDL Number MFCD00006427
Quantity 5 g
Molecular Weight (g/mol) 137.14
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.