Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse IgG3 Isotype Control

Catalog No. MG300
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MG300 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MG300 Supplier Invitrogen™ Supplier No. MG300
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Isotype Control

Mouse IgG3 Isotype Control for Flow

Mouse IgG3 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the least abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG3 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG3 is less commonly used in research and diagnostic applications compared to other IgG subtypes, but it is still used in some applications such as ELISA and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG3
Applications Flow Cytometry
Classification Monoclonal
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias IgG3; Ighg3; Immunoglobulin G; Immunoglobulin G3; Immunoglobulin heavy constant gamma 3; ImmunoglobulinG; ImmunoglobulinG3
Host Species Mouse
Purification Method Purified
Quantity 500 μL
Regulatory Status ASR
Whole Molecule Whole Antibody
Primary or Secondary Isotype Control (Primary)
Target Species Not Applicable
Content And Storage 2-8°C
Product Type Isotype Control
Form Liquid
Isotype IgG3
No. of Reactions 50 Tests
Shipping Condition Wet Ice
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.