Learn More
Uridine-5'-Diphosphoglucose Disodium Salt , MP Biomedicals™
Catalog No. MP210120883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210120883 250 mg
MP210120880 100 mg
MP210120890 500 mg
MP210120891 1 g
MP210120894 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP210120883 Supplier MP Biomedicals, Inc Supplier No. 0210120883
Add to Cart
Add to Cart
Uridine-5'-diphosphoglucose Disodium Salt is a biosynthetic product produced by the reaction of UTP and glucose-1-phosphate catalyzed by uridyl transferase.
  • Endogenous nucleotide sugar involved in glycosyltransferase reactions in metabolism
  • Shown to bind the P2Y14 receptor, an atypical P2Y receptor involved in the activation of dendritic cells and glial cells
  • Binds to and activates GPR17, inducing oligodendrocyte differentiation at a maximal concentration of 100μM
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov