Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Norclomipramine hydrochloride

Catalog No. N128010MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N128010MG 10 mg
N128025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N128010MG Supplier Sigma Aldrich Supplier No. N128010MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tricyclic antidepressant; an active metabolite of clomipramine that preferentially blocks the norepinephrine transporter.

Specifications

CAS 303-48-0
Molecular Formula C18H21ClN2 -+ HCl
Linear Formula C18H21ClN2 -+ HCl
MDL Number MFCD00792880
Quantity 10 mg
Synonym N-Desmethylclomipramine
Molecular Weight (g/mol) 337.29
Percent Purity ∼99% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.