Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Naphthoic acid

Catalog No. N190925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N190925G 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N190925G Supplier Sigma Aldrich Supplier No. N190925G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phenyl trifluoromethanesulfonate (phenyl triflate) is an aryl fluorosulphonate. it has been synthesized by the reaction of phenol with fluorosulphonic anhydride.phenyl trifluoromethanesulfonate (phenyl triflate) may be used in the following studies:
  • As an arylating agent for the asymmetric α-arylation of ketones catalyzed by pd(dba)2 and difluorphos
  • As a reactant in the one pot synthesis of carbazoles by palladium-catalyzed n-arylation of anilines with phenyl triflate followed by oxidative coupling
  • Synthesis of n-(2,6-diarylbenzoyl)anilines by diarylation of benzanilides with phenyl triflate in the presence of palladium-based catalyst
  • As an arylating agent in the synthesis of (r)-2-phenyl-2,3-dihydrofuran by the arylation of 2,3-dihydrofuran

Specifications

CAS 86-55-5
Melting Point 157°C to 160°C (lit.)
Boiling Point 300°C (lit.)
Molecular Formula C11H8O2
Linear Formula C10H7CO2H
MDL Number MFCD00004007
Quantity 5 mL
Synonym 1-Naphthalenecarboxylic acid
Molecular Weight (g/mol) 172.18
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.