Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich D-(+)-Neopterin

Catalog No. N338625MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N338625MG 100 g
N3386100MG 10 mg
N338610MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. N338625MG Supplier Sigma Aldrich Supplier No. N338625MG
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

D-(+)-Neopterin is a pteridine derivative, naturally found in Dictyostelium discoideum. It is found in significant amounts in the urine of cancer patients.D-(+)-Neopterin has been used as a NADPH oxidase inhibitor while assessing the ability of NADPH oxidases in breast cancer cells to produce reactive oxygen species (ROS). [NADPH= nicotinamide adenine dinucleotide phosphate]Pterin cofactor; precursor of BH4 (tetrahydrobiopterin).

Specifications

CAS 485-47-2
Melting Point 250°C
Molecular Formula C9H6O4
Linear Formula C9H6O4
MDL Number MFCD00003791
Quantity 100 g
Synonym D-erythro-Neopterin
Molecular Weight (g/mol) 178.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.