Learn More
BioVendor, LLC Endo-RS Endotoxin Recovery Kit, 1/Ea
SDP
Supplier:  BioVendor, LLC 609065
Encompass
Endo-RS is a novel sample preparation method enabling complete endotoxin recovery in biopharmaceutical drug formulations containing non-ionic surfactants such as Polysorbate 20, Polysorbate 80 or Octylphenol ethoxylates. Non-ionic surfactants are well known to cause Low Endotoxin Recovery (LER). Intensive studies of the phenomenon have shown that the physicochemical properties of surfactants render endotoxin molecules inaccessible (masked) for detection in such samples. The Endo-RS Endotoxin Recovery Kit provides the essential reagents and a detailed guideline for evaluating and establishing an optimized endotoxin demasking protocol for each individual formulation containing surfactants. Applying the Endo-RS Endotoxin Recovery Kit prior to endotoxin detection with the EndoLISA Endotoxin Detection Assay will ensure FDA-compliant and accurate results in spike validation studies and routine quality control testing.
Catalog No. NC0767903
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.