Learn More
SCANTIBODIES LABORATORY INC HBR 1, PURIFIED (3KC533) 1.0G
Supplier:  SCANTIBODIES LABORATORY INC 3KC5331G
Encompass
The 3KC533 Heterophilic Blocking Reagent 1, Purified from Scantibodies Laboratory, Inc. contains specific murine immunoglobulins that block the heterophilic interaction by active binding to the heterophilic antibodies, which are capable of cross-linking the capture and the detection antibodies used in the immunoassay, resulting in false positive readings. The attachment of HBR-1 to the heterophilic antibodies blocks this cross-linking, and eliminates the interference caused by the heterophilic antibodies in the humoral fluids. In addition to its active blocking, this product is also characteristic of its passive blockage of the heterophilic interaction as well. The HBR is a liquid reagent with a protein concentration of 20 ± 2 mg/ml. The immunoglobulins are dissolved in a phosphate buffer with a pH of 7.2-7.6. The immunoglobulins in this product are at a purity of greater than or equal to 95%.
Catalog No. NC0834272
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.