Learn More
SONY BIOTECHNOLOGY Beads, Instrument Setup and Maintenance
Supplier:  SONY BIOTECHNOLOGY LEB3001
Encompass
The Sony Biotechnology Cleaning Beads are polystyrene beads used to clean flow cells and fluidic systems in Sony cell sorters. They are designed to remove sample residue, prevent clogs, and maintain optimal instrument performance. The beads are 0.3-0.4mm in diameter and supplied in a 30mL bottle containing approximately 1.2 billion beads. To use, inject beads into the fluidics system and run for 1-2 minutes weekly or as needed. The beads can be reused up to 3 times if rinsed well between uses. This product helps maintain cell sorter fluidics for optimal sorting performance.
Catalog No. NC0876785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.