Learn More
                                    Fhc Inc FHC Inc Concentric Bipolar Microelectrode, Platinum-Iridium Inner, 75 mm Length
Supplier: Fhc Inc 30200
            The FHC Inc. Concentric Bipolar Electrode is a specialized tool for neuroscience research, designed for precise stimulation and multi-cell recording. Its robust construction, featuring a platinum-iridium inner pole and stainless steel outer pole, allows for effective stimulation in acute research settings, including *in vivo* and slice preparations. This electrode's rounded tip profile and optimal dimensions ensure large-field stimulation capabilities, making it ideal for researchers working with freely moving subjects.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.