Learn More
Omega Biotek Inc Total RNA Kit I, 200 preps, 1 x 10^7 eukaryotic cells or 30 mg tissue, up to 100 μg RNA yield, silica-based spin column, processing time less than 25 min, 40-70 μL elution volume
Supplier:  Omega Biotek Inc R683402
Encompass
Total RNA Kit I, 200 preps, 1 x 10^7 eukaryotic cells or 30 mg tissue, up to 100 μg RNA yield, silica-based spin column, processing time less than 25 min, 40-70 μL elution volume
Catalog No. NC0955262
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.