Learn More
SPEX CERTIPREP LLC 3P Stable Sulfide Standard, 1mg/mL
Supplier: SPEX CERTIPREP LLC 3P QCI147L
(NSI by Zeptometrix) An NSI Lab Solutions exclusive product. A sulfide CRM that is stable unopened for more than 2 years after manufacture. Precipitate free preservation with a proprietary buffer system and inert gas sealing. 1mg/mL Sulfide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.