Learn More
Sarstedt Inc Cell culture flask, T-25, surface: Suspension, Filter cap, 300/case
Supplier:  Sarstedt Inc 83.3910.502
Encompass_Preferred
Tissue culture flask T-25 has a canted neck and filter cap. These flasks are TC Tested (sterile, non-pyrogenic/endotoxin-free, non-cytotoxic, DNase-/RNase-/DNA-free).
Catalog No. NC1272994