Learn More
Iwai North America IMATRIX-511 1050UG 175UG X6
Supplier: Iwai North America N892012
iMatrix-511 is a recombinant Laminin- 511 E8 fragment expressed by CHO-cell. Laminin-511 E8 fragments allow ES/iPS cells and MSCs to be maintained in xeno-free culture conditions. It also enables the passaging of single cells and provides greater adhesion than full-length Laminin, Vitronectin, or Matrigel.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.