Learn More
BIOLIFE SOLUTIONS INC THAWSTART CFT2 AUTO CELL INSTR
SDP
Supplier:  BIOLIFE SOLUTIONS INC AST601
Encompass
  • Automated cell thawing instrument for cryopreserved samples.
  • Utilizes patent-pending STAR adaptive sensing technology.
  • Optimized for 1.8 - 2.0 mL cryogenic vials.
  • Typical thaw cycle time of 2-3 minutes.
  • Intuitive operation with audio and visual feedback.
  • Small footprint suitable for cell culture hoods.
  • Compatible with multiple storage conditions: liquid nitrogen, -80°C freezer, or dry ice.
  • Supports research or scalable commercial manufacturing applications.
  • Reproducible thawing performance promotes consistency and reduces operator error.
Catalog No. NC1575674
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.