Learn More
Hach Company BioTector B7000 Reagent
Supplier:  Hach Company 25255061
Encompass
BioTector B7000 Reagent 1.8 N Sulfuric Acid containing 80 mg/L Mn
Catalog No. NC1731400
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.