Learn More
Havels HAVELS No. 70 Autopsy Blade, Extra-Thick Carbon Steel, Compatible With #8 and #74 Handles, 100 Blades/Pack
Supplier: Havels AUT70
The HAVELS No. 70 Autopsy Blades are expertly crafted for precision and safety in dissection and autopsy contexts. Made of extra-thick non-sterile carbon steel, they feature a rounded tip to minimize puncture risk and come individually wrapped for hygiene. These blades are designed for long-lasting sharpness and compatibility with both #8 and #74 scalpel handles, making them an essential tool for pathology, post-mortem work, and research applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.