Learn More
Macherey-Nagel MACHEREY NUCLEODUR 100-3 C18 Ec, HPLC Column, 250 mm Length, 4.6 mm Diameter, 3 µm Particle Size
Supplier: Macherey-Nagel 760052.46
The NUCLEODUR 100-3 C18 ec 250x4.6mm from MACHEREY-NAGEL INC is a high-performance HPLC column designed for reversed-phase separations in both analytical and preparative applications. Its fully synthetic silica ensures excellent mechanical stability, while the C18 structure provides optimal selectivity for various compounds. It operates effectively across a pH range of 1 to 9 and supports temperatures up to 60 °C, making it ideal for routine analysis and method development.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.