Learn More
ASHLAND SPECIALTY INGREDIENTS Rad-Sure™ 15 Gy Irradiation Indicator, Visual Confirmation of Irradiation, FDA Cleared, Single-Use
Supplier: ASHLAND SPECIALTY INGREDIENTS 976631
The Rad-Sure™ 15 Gy Blood Irradiation Indicator is an essential quality control device that ensures blood products have been properly irradiated. Utilizing Gafchromic™ film technology, it provides a clear visual indication, changing from 'NOT IRRADIATED' to 'IRRADIATED' at a minimum dose of 15 Gy. This single-use indicator is FDA 510(k) cleared and adheres to AABB standards, offering reliable traceability with bar-coded lot numbers for enhanced compliance in blood banks.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.