Learn More
McKesson General Medical Alcare Hand Sanitizer, Antiseptic, Foaming, 62% Alcohol, 9 oz
Supplier:  McKesson General Medical 207082CS
Encompass_Preferred
Alcare Hand Sanitizer is a clinical-grade, foaming antiseptic handrub designed for rapid hand degerming in healthcare settings. It effectively eliminates pathogens like MRSA and VRE in just 5 seconds while moisturizing the skin for up to 2 hours. Its foaming mousse application ensures even coverage and minimal mess, setting it apart from conventional hand sanitizers. The product is also compatible with CHG, ensuring no interference with other antiseptic protocols, making it an essential addition to any healthcare hygiene routine.
Catalog No. NC2392090
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.