Learn More
Genesee Scientific Corporation 200 µL Premium Barrier Tips, RNase-Free, Pre-Sterilized, 200 µL
SDP
Supplier:  Genesee Scientific Corporation 23412
Encompass
200 µL Premium Barrier Tip: Ideal for precise liquid handling, these virgin polypropylene tips feature a barrier to prevent aerosol contamination. Certified free from biological contaminants, they ensure reliable results. Designed for universal compatibility with research-grade pipettors, they offer ergonomic benefits for effortless use and low-retentive characteristics for maximum sample recovery.
Catalog No. NC2400189
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.