Learn More
Air Science Usa Air Science USA GP HEPA Main Filter, High Efficiency, Captures 0.3 µm Particulates, Designed for Ductless Fume Hoods
SDP
Supplier:  Air Science Usa ASTM200001
Encompass_Preferred
The Air Science USA GP HEPA Main Filter provides high-efficiency particulate filtration to enhance laboratory safety by capturing airborne contaminants and fine particulates at 0.3 µm. It's designed for use in ductless fume hoods, integrating into the Multiplex™ Filtration System for superior protection against organic vapors and powders. This filter ensures optimal performance and reliability in maintaining a clean workspace.
Catalog No. NC2411047
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.