Learn More
Cygnus Technologies Cygnus Technologies CHO HCP 3G Assay Reagent Set, Semi-Quantitative Sandwich Immunoassay, Integrates with Gyrolab Systems, 24 Units
Supplier:  Cygnus Technologies G55015
Encompass
The Cygnus Technologies CHO HCP 3G Assay Reagent Set is a semi-quantitative solution for detecting Host Cell Protein (HCP) contamination in biopharmaceuticals derived from CHO cell lines. It integrates with Gyrolab systems, providing an automated workflow that enables high-throughput testing and reduces manual effort. Offering rapid results with high accuracy and broad detection, this set ensures reliable performance in regulatory scenarios, making it essential for purification process development and routine quality control.
Catalog No. NC2468297
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.