Learn More
Creative Bioarray M-07e human cell line, CSC-C0249, 1.0 mL volume, approximately 4 x 10^6 cells/ampoule, requires IL-3 or GM-CSF for growth, stored in liquid nitrogen
Supplier:  Creative Bioarray CSCC0249
Encompass
M-07e human cell line, CSC-C0249, 1.0 mL volume, approximately 4 x 10^6 cells/ampoule, requires IL-3 or GM-CSF for growth, stored in liquid nitrogen
Catalog No. NC2542037