Learn More
Cygnus Technologies Cygnus Technologies SP2/0 HCP ELISA Kit, High Sensitivity, Semi-Quantitative, 10
Supplier: Cygnus Technologies F1030
The Cygnus Technologies SP2/0 HCP ELISA Kit is a semi-quantitative assay designed for detecting host cell protein impurities in SP2/0 cell-derived products. It offers high sensitivity, with a limit of detection around 0.4 ng/mL and quantitation limit of 3 ng/mL. The kit's goat polyclonal antibodies provide 83% coverage against various HCP contaminants, ensuring reliable results for process development and routine final product testing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.