Learn More
Hach Company Ampule breaker for 20 mL glass ampules
Supplier:  Hach Company 8573700
Encompass
Ampule breaker designed to safely snap off the tips of 20 mL glass ampules, reducing the risk of contamination and injury when opening reagent ampules in laboratory testing.
  • Safely breaks 20 mL glass ampule tips.
  • Reduces contamination and injury risk when opening ampules.
  • Simple snap-action operation for fast use.
  • Compatible with standard 20 mL reagent ampules.
  • Replacement for older ampule breaker models.
Catalog No. NC2594283