Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human HLA-E*01:03 & B2M Monomer Protein, Peptide-Free, MALS-Verified, 25 μg

Supplier:  ACROBiosystems HLMH82EG25UG

Encompass_Preferred

Biotinylated human HLA-E*01:03 complexed with beta-2 microglobulin (B2M) is supplied as a peptide-free, MALS-verified monomer for tetramer assembly, receptor binding studies, and immunological assays. Supplied as 25 μg and expressed in HEK293 cells, the protein is biotinylated to enable streptavidin conjugation.

  • Biotinylated monomer format.
  • Contains HLA-E*01:03 (Gly22-Ile305) complexed with B2M.
  • Expression host: HEK293 (human).
  • Peptide free.
  • MALS verified for monodispersity.
  • Unit size: 25 μg.
  • Storage: -20 °C (recommended).
  • Applications: tetramer assembly, receptor binding, flow cytometry.

Catalog No. NC2821754


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.