Learn More
LAB ALLEY Lab Alley SDA 40B Ethanol 200 Proof (100%) Kosher Halal
SDP
Supplier:  LAB ALLEY EAS40B2004L
Encompass
This is a specially denatured alcohol formula 40B which contains trace amounts (less than 0.125% combined) of the denaturants tert-butyl alcohol and denatonium benzoate compliant with United States Title 27 of the Code of Federal Regulations Section 21.151. SDA-40B denatured ethanol is a popular choice for cosmetic applications because studies have shown low toxicity in topical applications. At Lab Alley, we understand that high quality reagents are essential to the integrity of your laboratory and manufacturing projects. That is why we believe you deserve to have full confidence in the quality of your reagents. Lab Alley provides high quality reliable SDA-40B ethanol ideal for your laboratory and manufacturing needs.
Catalog No. NC2828782
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.