Learn More
ACROBiosystems Human Glypican-3 (GPC3) R355A,R358A Fc-Tagged Recombinant Protein, 1 mg
Supplier: ACROBiosystems GP3H52551MG
Recombinant human Glypican-3 (GPC3) with R355A and R358A substitutions, expressed in HEK293 cells and fused to a C-terminal human IgG1 Fc tag. Supplied as 1 mg of highly purified protein for biochemical assays, antibody characterization, and assay development.
- Contains R355A and R358A substitutions.
- Amino acids Gln 25-His 559.
- Human IgG1 Fc tag at the C-terminus.
- Expressed in HEK293 cells.
- Purity >95% by SDS-PAGE; >90% by SEC-MALS.
- Calculated molecular weight 87.1 kDa; migrates higher due to glycosylation.
- Intended for assay development and antibody binding studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.