Learn More
ATCC ATCC 0.1% Gelatin Solution, 100 mL - For cell culture surface coating
Supplier: ATCC PCS999027
ATCC 0.1% Gelatin Solution is an aqueous 0.1% (w/v) gelatin preparation intended for coating cell culture vessels to promote cell attachment and adherence for routine cell culture applications.
- Promotes cell attachment on culture surfaces.
- Prepared at 0.1% (w/v) gelatin concentration.
- Provided as a 100 mL aqueous solution for laboratory use.
- Suitable for coating plates, flasks, and coverslips.
- Simple application: apply to surface and remove excess before seeding.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.