Learn More
Kam Controls Inc KAM Controls Inc 09063 Cathode Solution Cell, Electrolytic Iodine Generation, Diaphragm Type, 5 ml
Supplier: Kam Controls Inc 09063
The KAM CONTROLS INC Cathode Solution Cell is an essential component for accurate moisture measurement using coulometric Karl Fischer titration. It generates iodine electrolytically, making it suitable for analyzing water content in liquids like hydrocarbons and chemicals. This diaphragm-type cell ensures reliable results and is compatible with KAM Controls’ moisture analyzers, setting it apart in precision and application versatility.