Learn More
Phoenix Pharmaceuticals Inc Somatostatin-14 EIA Kit, Highly Sensitive Enzyme Immunoassay, 96 Wells
Supplier:  Phoenix Pharmaceuticals Inc EK06003
Encompass
The PHOENIX PHARMACEUTICALS INC SOMATOSTATIN-14 EIA Kit is a highly sensitive enzyme immunoassay designed for quantitative detection of Somatostatin-14 in various species. Its applications range from tumor biology research to insulin and glucose metabolism studies, making it a crucial tool for scientific research. Notably, this kit boasts a sensitivity of 0.13 ng/ml, a linear range of 0.13 – 1.16 ng/ml, and 100% cross-reactivity with several somatostatin forms, distinguishing it from other kits.
Catalog No. NC9867072
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.