Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyrithiamine hydrobromide

Catalog No. P02565MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02565MG 5 mg
P025610MG 10 mg
P02561MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P02565MG Supplier Sigma Aldrich Supplier No. P02565MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pyrithiamine hydrobromide

Specifications

CAS 534-64-5
Molecular Formula C14H18N4O -+ 2 HBr
Linear Formula C14H18N4O -+ 2 HBr
MDL Number MFCD00058193
Quantity 5 mg
Synonym 1-([4-Amino-2-methyl]-5-pyrimidylmethyl)-2-methyl-3-(beta-hydroxyethyl)pyridinium bromide; Neopyrithiamine
Molecular Weight (g/mol) 420.14
Percent Purity ∼95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.