Learn More
Description
- Involved in pharmacological studies investigating the use of flavonoids as inhibitors of cd38antioxidant and anthocyanidin:
- Found in various natural sources evaluated for inhibitory effects on colon and liver cancer cells
- Used as hyroperoxide and hydrogen peroxide scavenging substance
- Used to study relationship between structure, antioxidant capacity and redox potentials
- Studied to determine the mechanism of radical-scavengingantioxidant and anthocyanidin.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. p1659.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin
Specifications
Specifications
| CAS | 134-04-3 |
| Molecular Formula | C15H11ClO5 |
| Linear Formula | C15H11ClO5 |
| MDL Number | MFCD00017589 |
| Quantity | 10 mg |
| Synonym | 3,4 ,5,7-Tetrahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-1-benzopyrylium chloride; Pelargonidol chloride |
| Molecular Weight (g/mol) | 306.7 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.