Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenylacetyl chloride

Catalog No. P167535G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P167535G 5 g
P16753100G 100 g
P16753500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P167535G Supplier Sigma Aldrich Supplier No. P167535G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phenylacetyl chloride is used as a key precursor in the total synthesis of vialinin b
  • It is employed as a linker to prepare dendrimers and also used in the synthesis of various conjugated aromatic small molecules

Specifications

CAS 103-80-0
Density 1.169 g/mL (at 25°C (literature))
Boiling Point 94°C to 95°C (12 mmHg)
Molecular Formula C8H7ClO
Refractive Index n20/D 1.5325 (literature)
Linear Formula C6H5CH2COCl
MDL Number MFCD00000729
Quantity 5 g
Synonym alpha-Toluyl chloride
Molecular Weight (g/mol) 154.59
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.